ENA308584756 Enamine

4-chloro-2-[(cyclopropylmethyl)amino]benzonitrile

  • MDL number MFCD12119002
  • Cas Number : 1153005-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69128
Availability details for
Quantity
  • Availability
  • Add to Cart