ENA308584758 Enamine

4-ethyl-N-[1-(4-fluorophenyl)ethyl]cyclohexan-1-amine

  • MDL number MFCD11117056
  • Cas Number : 1132655-07-2
  • Molecular Weight : 249.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69599
Availability details for
Quantity
  • Availability
  • Add to Cart