ENA308584759 Enamine

2-(2-{[2-(dimethylamino)ethyl]amino}ethoxy)ethan-1-ol

  • MDL number MFCD12783673
  • Cas Number : 1250107-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69600
Availability details for
Quantity
  • Availability
  • Add to Cart