ENA308584761 Enamine

2-methyl-4-{[1,2,4]triazolo[4,3-a]pyridin-3-yl}aniline

  • MDL number MFCD09944785
  • Cas Number : 1016839-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69129
Availability details for
Quantity
  • Availability
  • Add to Cart