ENA308584764 Enamine

4-[(pyridin-4-yl)methoxy]aniline

  • MDL number MFCD04971041
  • Cas Number : 105350-42-3
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69606
Availability details for
Quantity
  • Availability
  • Add to Cart