ENA308584766 Enamine

1-benzyl-2,2-dimethylpiperazine

  • MDL number MFCD11847783
  • Cas Number : 846052-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69608
Availability details for
Quantity
  • Availability
  • Add to Cart