ENA308584767 Enamine

3-[(2-methoxyethyl)amino]propanenitrile

  • MDL number MFCD10687307
  • Cas Number : 35855-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69134
Availability details for
Quantity
  • Availability
  • Add to Cart