ENA308584774 Enamine

3-[ethyl(methyl)carbamoyl]benzoic acid

  • MDL number MFCD12173497
  • Cas Number : 1010797-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69627
Availability details for
Quantity
  • Availability
  • Add to Cart