ENA308584775 Enamine

propan-2-yl 2-aminoacetate hydrochloride

  • MDL number MFCD00151826
  • Cas Number : 14019-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69629

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69629
Availability details for
Quantity
  • Availability
  • Add to Cart