ENA308584785 Enamine

(3-phenylpropyl)hydrazine hydrochloride

  • MDL number MFCD02322594
  • Cas Number : 24214-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69640
Availability details for
Quantity
  • Availability
  • Add to Cart