ENA308584799 Enamine

quinoline-4-sulfonic acid

  • MDL number MFCD03426331
  • Cas Number : 6046-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 317...319
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69141

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69141
Availability details for
Quantity
  • Availability
  • Add to Cart