ENA308584802 Enamine

2-amino-1-(pyrrolidin-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD07366742
  • Cas Number : 35855-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69144

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69144
Availability details for
Quantity
  • Availability
  • Add to Cart