ENA308584803 Enamine

3-(2,4-difluorobenzoyl)pyridine

  • MDL number MFCD11105482
  • Cas Number : 133385-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69664
Availability details for
Quantity
  • Availability
  • Add to Cart