ENA308584807 Enamine

3-methoxybenzene-1-carboximidamide hydrochloride

  • MDL number MFCD04114430
  • Cas Number : 26113-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-165
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69670
Availability details for
Quantity
  • Availability
  • Add to Cart