ENA308584813 Enamine

5H,6H,7H,8H-imidazo[1,2-a]pyrimidine

  • MDL number MFCD08273903
  • Cas Number : 67139-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-109
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69679
Availability details for
Quantity
  • Availability
  • Add to Cart