ENA308584814 Enamine

3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylic acid

  • MDL number MFCD11878407
  • Cas Number : 208772-72-9
  • Molecular Weight : 193.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 302-305
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69680

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-69680
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart