ENA308584819 Enamine

5,5-dimethylhexan-1-amine

  • MDL number MFCD09913598
  • Cas Number : 60996-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69686

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69686
Availability details for
Quantity
  • Availability
  • Add to Cart