ENA308584826 Enamine

4-[methyl(propan-2-yl)amino]butan-2-one

  • MDL number MFCD11190951
  • Cas Number : 5843-24-3
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69152
Availability details for
Quantity
  • Availability
  • Add to Cart