ENA308584831 Enamine

1,4-diphenylbutan-2-one

  • MDL number MFCD05149240
  • Cas Number : 37985-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69718
Availability details for
Quantity
  • Availability
  • Add to Cart