ENA308584840 Enamine

3-{[(pyridin-2-yl)methyl]amino}propanenitrile

  • MDL number MFCD11166782
  • Cas Number : 112086-58-5
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69156
Availability details for
Quantity
  • Availability
  • Add to Cart