ENA308584841 Enamine

2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylic acid

  • MDL number MFCD16631264
  • Cas Number : 1195159-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69735
Availability details for
Quantity
  • Availability
  • Add to Cart