ENA308584845 Enamine

1-(4-chlorophenyl)-1-cyclopentylmethanamine

  • MDL number MFCD11127103
  • Cas Number : 1855-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69739
Availability details for
Quantity
  • Availability
  • Add to Cart