ENA308584851 Enamine

1-(2-methylbenzoyl)piperidin-4-one

  • MDL number MFCD09945852
  • Cas Number : 203186-44-1
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69756
Availability details for
Quantity
  • Availability
  • Add to Cart