ENA308584854 Enamine

N-(4,5-dihydro-1,3-thiazol-2-yl)pyridin-3-amine

  • MDL number MFCD11202404
  • Cas Number : 112934-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69157
Availability details for
Quantity
  • Availability
  • Add to Cart