ENA308584856 Enamine

3-(3-methylfuran-2-yl)-3-oxopropanenitrile

  • MDL number MFCD09929195
  • Cas Number : 158386-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69761
Availability details for
Quantity
  • Availability
  • Add to Cart