ENA308584858 Enamine

6-(dimethylcarbamoyl)-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD12173513
  • Cas Number : 1154955-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136...138
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69764

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69764
Availability details for
Quantity
  • Availability
  • Add to Cart