ENA308584862 Enamine

4-[(2-chlorophenyl)methoxy]benzene-1-sulfonamide

  • MDL number MFCD12568007
  • Cas Number : 1096973-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69768
Availability details for
Quantity
  • Availability
  • Add to Cart