ENA308584863 Enamine

2-fluoro-5-[(2-methyl-2,3-dihydro-1H-indol-1-yl)sulfonyl]benzoic acid

  • MDL number MFCD10607506
  • Cas Number : 1030149-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69769
Availability details for
Quantity
  • Availability
  • Add to Cart