ENA308584864 Enamine

2-{[(benzyloxy)carbonyl]amino}-2-methylpentanoic acid

  • MDL number MFCD12689596
  • Cas Number : 1155991-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69770
Availability details for
Quantity
  • Availability
  • Add to Cart