ENA308584865 Enamine

[1-(2,4-dichlorophenyl)ethyl]thiourea

  • MDL number MFCD10688722
  • Cas Number : 1152515-60-0
  • Molecular Weight : 249.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69771
Availability details for
Quantity
  • Availability
  • Add to Cart