ENA308584866 Enamine

3-(chloromethyl)-5-(furan-3-yl)-1,2,4-oxadiazole

  • MDL number MFCD10695999
  • Cas Number : 1056157-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69772
Availability details for
Quantity
  • Availability
  • Add to Cart