ENA308584869 Enamine

(1,2,3,4-tetrahydronaphthalen-1-yl)methanol

  • MDL number MFCD16857988
  • Cas Number : 66377-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69779
Availability details for
Quantity
  • Availability
  • Add to Cart