ENA308584871 Enamine

1-cyclopropyl-2-phenylethan-1-one

  • MDL number MFCD11934440
  • Cas Number : 14113-94-1
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69781
Availability details for
Quantity
  • Availability
  • Add to Cart