ENA308584878 Enamine

1-(2,5-dichlorophenyl)-2-(4-methoxyphenyl)ethan-1-one

  • MDL number MFCD12569776
  • Cas Number : 1098349-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69790
Availability details for
Quantity
  • Availability
  • Add to Cart