ENA308584879 Enamine

tert-butyl 2-[(2-methoxyethyl)amino]acetate

  • MDL number MFCD12148598
  • Cas Number : 66116-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69791
Availability details for
Quantity
  • Availability
  • Add to Cart