ENA308584881 Enamine

2-oxo-6-propyl-1,2,5,6,7,8-hexahydroquinoline-3-carbonitrile

  • MDL number MFCD16070456
  • Cas Number : 1248123-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69794
Availability details for
Quantity
  • Availability
  • Add to Cart