ENA308584889 Enamine

1-(prop-2-enoyl)piperidine-2-carboxylic acid

  • MDL number MFCD09948915
  • Cas Number : 1163678-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69809
Availability details for
Quantity
  • Availability
  • Add to Cart