ENA308584890 Enamine

1-methyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00661384
  • Cas Number : 15506-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-184
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69811
Availability details for
Quantity
  • Availability
  • Add to Cart