ENA308584894 Enamine

3-[2-(difluoromethoxy)phenyl]-1,2-oxazol-5-amine

  • MDL number MFCD11156292
  • Cas Number : 1021245-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69820

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-69820
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart