ENA308584896 Enamine

1-(4-amino-1H-pyrazol-1-yl)-3-(morpholin-4-yl)propan-2-ol

  • MDL number MFCD11654972
  • Cas Number : 1152624-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69827
Availability details for
Quantity
  • Availability
  • Add to Cart