ENA308584898 Enamine

tert-butyl 6-amino-1,2,3,4-tetrahydroquinoline-1-carboxylate

  • MDL number MFCD10566524
  • Cas Number : 1152923-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69829

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-69829
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart