ENA308584904 Enamine

1-[2-(4-fluorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-amine

  • MDL number MFCD14615036
  • Cas Number : 1250123-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69835
Availability details for
Quantity
  • Availability
  • Add to Cart