ENA308584908 Enamine

6-amino-1-methyl-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD11987121
  • Cas Number : 233775-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101...103
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69848

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-69848
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart