ENA308584910 Enamine

7-chloro-2,3-dihydro-1-benzofuran-3-one

  • MDL number MFCD08544412
  • Cas Number : 3260-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69850
Availability details for
Quantity
  • Availability
  • Add to Cart