ENA308584911 Enamine

2-(hydroxymethyl)benzamide

  • MDL number MFCD09834506
  • Cas Number : 33832-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69851
Availability details for
Quantity
  • Availability
  • Add to Cart