ENA308584913 Enamine

1-[4-(thiophen-2-yl)-1,3-thiazol-2-yl]piperazine

  • MDL number MFCD11592088
  • Cas Number : 1082363-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69853

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69853
Availability details for
Quantity
  • Availability
  • Add to Cart