ENA308584914 Enamine

1-(1-cyclopropylethyl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD12195218
  • Cas Number : 1153367-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69854
Availability details for
Quantity
  • Availability
  • Add to Cart