ENA308584919 Enamine

4-(5-chloro-1,3-benzothiazol-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD12169542
  • Cas Number : 1187636-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 274...276
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69860

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69860
Availability details for
Quantity
  • Availability
  • Add to Cart