ENA308584921 Enamine

3-(2-fluorophenyl)pyrrolidine-2,5-dione

  • MDL number MFCD16303588
  • Cas Number : 1225665-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69863
Availability details for
Quantity
  • Availability
  • Add to Cart