ENA308584922 Enamine

3-(cyclopentyloxy)-2-methylaniline

  • MDL number MFCD12170012
  • Cas Number : 1154909-64-4
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69865
Availability details for
Quantity
  • Availability
  • Add to Cart